The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152546.2 | NP_689759.2 | 7 - 8 | 1177 | 144 |
GenBank mRNA | AK058015.1 | - | 7 - 8 | 1177 | 144 |
AY611630.2 | - | 7 - 8 | 1105 | 144 | |
BC017102.1 | - | 2 - 3 | 760 | 144 | |
BC031222.1 | - | 7 - 8 | 1148 | 144 |