Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC4A10
Entrez Gene ID
57282
Gene Name
solute carrier family 4 member 10
Gene Aliases
NBCn2, NCBE
Chromosome Location
Chr.2: 161624335 - 161985276 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178015.1 NP_001171486.1 12 - 13 1628 113
NM_001178016.1 NP_001171487.1 12 - 13 1674 113
NM_022058.3 NP_071341.2 11 - 12 1538 113
XM_005246694.4 XP_005246751.1 13 - 14 1683 113
XM_005246695.4 XP_005246752.1 12 - 13 1593 113
XM_011511508.2 XP_011509810.1 12 - 13 1471 113
XM_011511509.2 XP_011509811.1 12 - 13 1468 113
XM_011511510.2 XP_011509812.1 12 - 13 1471 113
XM_011511511.2 XP_011509813.1 11 - 12 1381 113
XM_011511512.2 XP_011509814.1 11 - 12 1378 113
XM_011511513.2 XP_011509815.1 15 - 16 1960 113
XM_017004541.1 XP_016860030.1 12 - 13 1468 113
XM_017004542.1 XP_016860031.1 11 - 12 1378 113
XM_017004543.1 XP_016860032.1 11 - 12 1381 113
XM_017004544.1 XP_016860033.1 11 - 12 1378 113
XM_017004545.1 XP_016860034.1 14 - 15 1869 113
XM_017004546.1 XP_016860035.1 14 - 15 1870 113
XM_017004547.1 XP_016860036.1 13 - 14 1683 113
XM_017004548.1 XP_016860037.1 13 - 14 1657 113
XM_017004549.1 XP_016860038.1 13 - 14 1745 113
XM_017004550.1 XP_016860039.1 12 - 13 1590 113
XM_017004551.1 XP_016860040.1 8 - 9 966 113
XM_017004552.1 XP_016860041.1 12 - 13 1487 113
XM_017004553.1 XP_016860042.1 12 - 13 1487 113
XM_017004554.1 XP_016860043.1 2 - 3 670 113
GenBank mRNA AB040457.1 - 11 - 12 1446 113
AK293793.1 - 12 - 13 1587 113
AY376402.1 - 12 - 13 1444 113
BC136269.1 - 11 - 12 1424 113
BC143714.1 - 12 - 13 1514 113

View Full Product Details