Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SLC4A10
- Entrez Gene ID
-
57282
- Gene Name
- solute carrier family 4 member 10
- Gene Aliases
-
NBCn2, NCBE
- Chromosome Location
-
Chr.2: 161624335 - 161985276 on Build GRCh38
View Full Product Details