The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015715.4 | NP_056530.2 | 3 - 4 | 1035 | 106 |
XM_011530204.1 | XP_011528506.1 | 3 - 4 | 1257 | 106 | |
XM_011530205.1 | XP_011528507.1 | 3 - 4 | 1246 | 106 | |
GenBank mRNA | AF220490.1 | - | 3 - 4 | 1035 | 106 |
BC025316.1 | - | 3 - 4 | 911 | 106 |