The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015715.4 | NP_056530.2 | 6 - 7 | 1569 | 91 |
XM_011530204.1 | XP_011528506.1 | 6 - 7 | 1791 | 91 | |
XM_011530205.1 | XP_011528507.1 | 6 - 7 | 1780 | 91 | |
GenBank mRNA | AF220490.1 | - | 6 - 7 | 1569 | 91 |
BC025316.1 | - | 6 - 7 | 1445 | 91 |