The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014716.3 | NP_055531.1 | 2 - 3 | 320 | 80 |
GenBank mRNA | BC018543.1 | - | 2 - 3 | 295 | 80 |
BT009788.1 | - | 2 - 3 | 114 | 80 | |
D30758.1 | - | 2 - 3 | 302 | 80 |