The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001608.3 | NP_001599.1 | 6 - 7 | 997 | 134 |
XM_005246517.4 | XP_005246574.1 | 6 - 7 | 1449 | 134 | |
XM_017003955.1 | XP_016859444.1 | 6 - 7 | 1425 | 134 | |
GenBank mRNA | AK313498.1 | - | 6 - 7 | 951 | 134 |
BC039063.1 | - | 6 - 7 | 942 | 134 | |
BC064549.1 | - | 6 - 7 | 929 | 134 | |
M74096.1 | - | 6 - 7 | 774 | 134 |