The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012089.2 | NP_036221.2 | 12 - 13 | 2031 | 105 |
XM_011544135.2 | XP_011542437.1 | 12 - 13 | 2802 | 105 | |
XM_011544136.2 | XP_011542438.1 | 10 - 11 | 1268 | 105 | |
GenBank mRNA | AB013380.1 | - | 12 - 13 | 2018 | 105 |
AF216833.1 | - | 12 - 13 | 2031 | 105 | |
AK172794.1 | - | 11 - 12 | 1287 | 105 | |
BC064930.1 | - | 12 - 13 | 2026 | 105 | |
DQ786241.1 | - | 5 - 6 | 409 | 105 |