The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013233.2 | NP_037365.2 | 13 - 14 | 1468 | 90 |
XM_017003813.1 | XP_016859302.1 | 13 - 14 | 1414 | 90 | |
XM_017003814.1 | XP_016859303.1 | 13 - 14 | 1414 | 90 | |
XM_017003815.1 | XP_016859304.1 | 13 - 14 | 2754 | 90 | |
XM_017003817.1 | XP_016859306.1 | 10 - 11 | 827 | 90 | |
GenBank mRNA | AF017635.1 | - | 7 - 8 | 500 | 90 |
AF030403.1 | - | 13 - 14 | 1560 | 90 | |
AF099989.1 | - | 13 - 14 | 1487 | 90 | |
BX647113.1 | - | 4 - 5 | 409 | 90 | |
KJ534948.1 | - | 14 - 15 | 1251 | 90 | |
KJ534949.1 | - | 14 - 15 | 1251 | 90 | |
KJ534951.1 | - | 14 - 15 | 1242 | 90 | |
KJ535059.1 | - | 14 - 15 | 1231 | 90 |