The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013233.2 | NP_037365.2 | 15 - 16 | 1590 | 116 |
XM_005246465.2 | XP_005246522.1 | 14 - 15 | 1473 | 116 | |
XM_017003813.1 | XP_016859302.1 | 15 - 16 | 1536 | 116 | |
XM_017003815.1 | XP_016859304.1 | 15 - 16 | 2876 | 116 | |
XM_017003817.1 | XP_016859306.1 | 12 - 13 | 949 | 116 | |
GenBank mRNA | AF017635.1 | - | 9 - 10 | 622 | 116 |
AF030403.1 | - | 15 - 16 | 1682 | 116 | |
AF099989.1 | - | 15 - 16 | 1609 | 116 | |
BX647113.1 | - | 6 - 7 | 531 | 116 | |
KJ534948.1 | - | 16 - 17 | 1373 | 116 | |
KJ534949.1 | - | 16 - 17 | 1373 | 116 | |
KJ534950.1 | - | 14 - 15 | 1220 | 116 | |
KJ534951.1 | - | 16 - 17 | 1364 | 116 | |
KJ535059.1 | - | 16 - 17 | 1353 | 116 |