The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013233.2 | NP_037365.2 | 6 - 7 | 895 | 64 |
XM_005246465.2 | XP_005246522.1 | 6 - 7 | 841 | 64 | |
XM_017003813.1 | XP_016859302.1 | 6 - 7 | 841 | 64 | |
XM_017003814.1 | XP_016859303.1 | 6 - 7 | 841 | 64 | |
XM_017003815.1 | XP_016859304.1 | 6 - 7 | 2181 | 64 | |
XM_017003816.1 | XP_016859305.1 | 6 - 7 | 841 | 64 | |
XM_017003817.1 | XP_016859306.1 | 3 - 4 | 254 | 64 | |
GenBank mRNA | AF030403.1 | - | 6 - 7 | 987 | 64 |
AF099989.1 | - | 6 - 7 | 914 | 64 | |
KJ534948.1 | - | 7 - 8 | 678 | 64 | |
KJ534949.1 | - | 7 - 8 | 678 | 64 | |
KJ534951.1 | - | 7 - 8 | 669 | 64 | |
KJ535059.1 | - | 7 - 8 | 658 | 64 |