The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008405.2 | NP_001008405.1 | 7 - 8 | 778 | 143 |
XM_006716051.2 | XP_006716114.1 | 8 - 9 | 1099 | 143 | |
XM_011516406.2 | XP_011514708.1 | 8 - 9 | 862 | 143 | |
GenBank mRNA | BG818947.1 | - | 3 - 4 | 294 | 143 |
BP261060.1 | - | 5 - 6 | 442 | 143 |