The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177506.1 | NP_001170977.1 | 10 - 11 | 1153 | 124 |
NM_001177507.1 | NP_001170978.1 | 9 - 10 | 1057 | 124 | |
NM_001637.3 | NP_001628.1 | 10 - 11 | 1153 | 124 | |
XM_011515333.2 | XP_011513635.1 | 10 - 11 | 1098 | 124 | |
XM_011515334.2 | XP_011513636.1 | 9 - 10 | 1052 | 124 | |
XM_011515335.2 | XP_011513637.1 | 11 - 12 | 1162 | 124 | |
XM_011515336.2 | XP_011513638.1 | 10 - 11 | 1098 | 124 | |
XM_011515338.2 | XP_011513640.1 | 10 - 11 | 1098 | 124 | |
XM_011515339.2 | XP_011513641.1 | 10 - 11 | 1098 | 124 | |
XM_011515340.2 | XP_011513642.1 | 10 - 11 | 1098 | 124 | |
XM_011515341.2 | XP_011513643.1 | 10 - 11 | 1098 | 124 | |
XM_017012102.1 | XP_016867591.1 | 10 - 11 | 1098 | 124 | |
XM_017012103.1 | XP_016867592.1 | 7 - 8 | 429 | 124 | |
XM_017012104.1 | XP_016867593.1 | 10 - 11 | 1098 | 124 | |
XM_017012106.1 | XP_016867595.1 | 10 - 11 | 1098 | 124 | |
GenBank mRNA | AK297016.1 | - | 9 - 10 | 943 | 124 |
AK308215.1 | - | 10 - 11 | 1054 | 124 | |
BC025698.1 | - | 10 - 11 | 1058 | 124 | |
M62840.1 | - | 10 - 11 | 1026 | 124 |