Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AOAH
Entrez Gene ID
313
Gene Name
acyloxyacyl hydrolase
Gene Aliases
-
Chromosome Location
Chr.7: 36512042 - 36724549 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177506.1 NP_001170977.1 16 - 17 1708 91
NM_001177507.1 NP_001170978.1 15 - 16 1612 91
NM_001637.3 NP_001628.1 16 - 17 1708 91
XM_011515333.2 XP_011513635.1 15 - 16 1578 91
XM_011515334.2 XP_011513636.1 15 - 16 1607 91
XM_011515335.2 XP_011513637.1 17 - 18 1717 91
XM_011515336.2 XP_011513638.1 16 - 17 1653 91
XM_011515338.2 XP_011513640.1 16 - 17 1653 91
XM_011515339.2 XP_011513641.1 16 - 17 1653 91
XM_011515340.2 XP_011513642.1 16 - 17 1653 91
XM_011515342.2 XP_011513644.1 6 - 7 608 91
XM_017012102.1 XP_016867591.1 15 - 16 1578 91
XM_017012103.1 XP_016867592.1 13 - 14 984 91
XM_017012104.1 XP_016867593.1 15 - 16 1578 91
XM_017012105.1 XP_016867594.1 7 - 8 768 91
GenBank mRNA AK223476.1 - 16 - 17 1624 91
AK297016.1 - 15 - 16 1498 91
AK298644.1 - 6 - 7 529 91
AK308215.1 - 16 - 17 1609 91
BC025698.1 - 16 - 17 1613 91
M62840.1 - 16 - 17 1581 91

View Full Product Details