The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177506.1 | NP_001170977.1 | 19 - 20 | 1924 | 123 |
NM_001177507.1 | NP_001170978.1 | 18 - 19 | 1828 | 123 | |
NM_001637.3 | NP_001628.1 | 19 - 20 | 1924 | 123 | |
XM_011515333.2 | XP_011513635.1 | 18 - 19 | 1794 | 123 | |
XM_011515334.2 | XP_011513636.1 | 18 - 19 | 1823 | 123 | |
XM_011515335.2 | XP_011513637.1 | 20 - 21 | 1933 | 123 | |
XM_011515336.2 | XP_011513638.1 | 19 - 20 | 1869 | 123 | |
XM_011515338.2 | XP_011513640.1 | 19 - 20 | 1869 | 123 | |
XM_011515342.2 | XP_011513644.1 | 9 - 10 | 824 | 123 | |
XM_017012102.1 | XP_016867591.1 | 18 - 19 | 1794 | 123 | |
XM_017012103.1 | XP_016867592.1 | 16 - 17 | 1200 | 123 | |
XM_017012104.1 | XP_016867593.1 | 18 - 19 | 1794 | 123 | |
XM_017012105.1 | XP_016867594.1 | 10 - 11 | 984 | 123 | |
GenBank mRNA | AK223476.1 | - | 19 - 20 | 1840 | 123 |
AK297016.1 | - | 18 - 19 | 1714 | 123 | |
AK298644.1 | - | 9 - 10 | 745 | 123 | |
AK308215.1 | - | 19 - 20 | 1825 | 123 | |
BC025698.1 | - | 19 - 20 | 1829 | 123 | |
M62840.1 | - | 19 - 20 | 1797 | 123 |