The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297644.1 | NP_001284573.1 | 5 - 6 | 722 | 123 |
NM_001297645.1 | NP_001284574.1 | 5 - 6 | 722 | 123 | |
NM_001318000.1 | NP_001304929.1 | 5 - 6 | 722 | 123 | |
NM_005869.3 | NP_005860.2 | 5 - 6 | 722 | 123 | |
XM_011543095.2 | XP_011541397.2 | 5 - 6 | 691 | 123 | |
XM_011543096.1 | XP_011541398.1 | 5 - 6 | 691 | 123 | |
XM_017008943.1 | XP_016864432.1 | 5 - 6 | 691 | 123 | |
GenBank mRNA | AF039692.1 | - | 5 - 6 | 674 | 123 |
AF039693.1 | - | 1 - 2 | 113 | 123 | |
AK225105.1 | - | 5 - 6 | 689 | 123 | |
AK225161.1 | - | 5 - 6 | 694 | 123 | |
AK308223.1 | - | 5 - 6 | 691 | 123 | |
AK312954.1 | - | 5 - 6 | 691 | 123 | |
AY358569.1 | - | 5 - 6 | 683 | 123 | |
BC012117.1 | - | 5 - 6 | 675 | 123 | |
BX647971.1 | - | 5 - 6 | 723 | 123 |