The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032286.2 | NP_115662.2 | 2 - 3 | 251 | 96 |
GenBank mRNA | AY277599.1 | - | 2 - 3 | 208 | 96 |
AY453397.1 | - | 2 - 3 | 249 | 96 | |
AY598325.1 | - | 2 - 3 | 208 | 96 | |
BC003353.1 | - | 2 - 3 | 251 | 96 | |
BI597082.1 | - | 2 - 3 | 240 | 96 |