The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114377.1 | NP_001107849.1 | 2 - 3 | 401 | 55 |
XM_011543916.2 | XP_011542218.1 | 3 - 4 | 1931 | 55 | |
XM_017029566.1 | XP_016885055.1 | 3 - 4 | 1931 | 55 | |
XM_017029567.1 | XP_016885056.1 | 2 - 3 | 326 | 55 | |
GenBank mRNA | AJ005891.1 | - | 2 - 3 | 168 | 55 |
AK299988.1 | - | 2 - 3 | 285 | 55 | |
BC143785.1 | - | 1 - 2 | 243 | 55 | |
DQ010327.1 | - | 1 - 2 | 213 | 55 |