The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114377.1 | NP_001107849.1 | 5 - 6 | 735 | 71 |
NM_014009.3 | NP_054728.2 | 6 - 7 | 840 | 71 | |
XM_006724533.2 | XP_006724596.2 | 5 - 6 | 1015 | 71 | |
XM_011543916.2 | XP_011542218.1 | 6 - 7 | 2265 | 71 | |
XM_017029565.1 | XP_016885054.1 | 7 - 8 | 2301 | 71 | |
XM_017029566.1 | XP_016885055.1 | 6 - 7 | 2265 | 71 | |
XM_017029567.1 | XP_016885056.1 | 5 - 6 | 660 | 71 | |
GenBank mRNA | AF277993.1 | - | 6 - 7 | 840 | 71 |
AJ005891.1 | - | 5 - 6 | 502 | 71 | |
AK292052.1 | - | 6 - 7 | 724 | 71 | |
AK299988.1 | - | 5 - 6 | 619 | 71 | |
BC113401.1 | - | 5 - 6 | 682 | 71 | |
BC113403.1 | - | 5 - 6 | 682 | 71 | |
BC143785.1 | - | 4 - 5 | 577 | 71 | |
BC143786.1 | - | 4 - 5 | 751 | 71 | |
DQ010327.1 | - | 4 - 5 | 547 | 71 | |
EF534714.1 | - | 5 - 6 | 652 | 71 | |
EU855812.1 | - | 5 - 6 | 652 | 71 |