Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FOXP3
Entrez Gene ID
50943
Gene Name
forkhead box P3
Gene Aliases
AIID, DIETER, IPEX, JM2, PIDX, XPID
Chromosome Location
Chr.X: 49250436 - 49266505 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114377.1 NP_001107849.1 6 - 7 820 66
NM_014009.3 NP_054728.2 7 - 8 925 66
XM_006724533.2 XP_006724596.2 6 - 7 1100 66
XM_011543916.2 XP_011542218.1 7 - 8 2350 66
XM_017029565.1 XP_016885054.1 8 - 9 2386 66
XM_017029566.1 XP_016885055.1 7 - 8 2350 66
XM_017029567.1 XP_016885056.1 6 - 7 745 66
GenBank mRNA AF277993.1 - 7 - 8 925 66
AJ005891.1 - 6 - 7 587 66
AK292052.1 - 7 - 8 809 66
BC113401.1 - 6 - 7 767 66
BC113403.1 - 6 - 7 767 66
BC143785.1 - 5 - 6 662 66
BC143786.1 - 5 - 6 836 66
DQ010327.1 - 5 - 6 632 66
EF534714.1 - 6 - 7 737 66

View Full Product Details