The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003076.4 | NP_003067.3 | 7 - 8 | 1049 | 76 |
NM_139071.2 | NP_620710.2 | 7 - 8 | 1049 | 76 | |
XM_005269107.3 | XP_005269164.2 | 6 - 7 | 926 | 76 | |
GenBank mRNA | AF109733.1 | - | 7 - 8 | 837 | 76 |
AK130517.1 | - | 8 - 9 | 789 | 76 | |
AK292700.1 | - | 7 - 8 | 713 | 76 | |
BC009368.2 | - | 7 - 8 | 999 | 76 | |
CN272379.1 | - | 6 - 7 | 579 | 76 | |
DA825057.1 | - | 3 - 4 | 320 | 76 | |
U66617.1 | - | 7 - 8 | 1027 | 76 |