The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127229.1 | NP_001120701.1 | 2 - 3 | 294 | 112 |
NM_001127230.1 | NP_001120702.1 | 2 - 3 | 198 | 112 | |
NM_017900.2 | NP_060370.1 | 2 - 3 | 375 | 112 | |
GenBank mRNA | AK000615.1 | - | 2 - 3 | 137 | 112 |
BC022808.1 | - | 1 - 2 | 166 | 112 | |
BC062333.1 | - | 2 - 3 | 140 | 112 | |
BE903314.1 | - | 2 - 3 | 183 | 112 | |
BF664406.1 | - | 2 - 3 | 111 | ||
BG764236.1 | - | 2 - 3 | 247 | 112 | |
BM781908.1 | - | 2 - 3 | 294 | 112 | |
BP198074.1 | - | 2 - 3 | 198 | 112 | |
DN994953.1 | - | 2 - 3 | 188 | 112 |