The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193268.1 | NP_001180197.1 | 19 - 20 | 2270 | 77 |
NM_001193269.1 | NP_001180198.1 | 19 - 20 | 2174 | 77 | |
NM_012155.2 | NP_036287.1 | 16 - 17 | 1635 | 77 | |
NR_034098.1 | 14 - 15 | 1355 | 77 | ||
XM_006723125.2 | XP_006723188.1 | 19 - 20 | 2351 | 77 | |
XM_006723126.2 | XP_006723189.1 | 19 - 20 | 2159 | 77 | |
XM_011526694.2 | XP_011524996.1 | 18 - 19 | 2258 | 77 | |
XM_011526695.2 | XP_011524997.1 | 18 - 19 | 2219 | 77 | |
XM_011526698.2 | XP_011525000.1 | 16 - 17 | 1677 | 77 | |
XM_011526700.2 | XP_011525002.1 | 16 - 17 | 1623 | 77 | |
XM_011526701.2 | XP_011525003.1 | 11 - 12 | 1226 | 77 | |
XM_017026550.1 | XP_016882039.1 | 15 - 16 | 1519 | 77 | |
GenBank mRNA | AB209773.1 | - | 13 - 14 | 1371 | 77 |
AF103939.1 | - | 16 - 17 | 1629 | 77 | |
AK026616.1 | - | 14 - 15 | 1333 | 77 | |
AK225043.1 | - | 16 - 17 | 1621 | 77 | |
AK294668.1 | - | 19 - 20 | 2092 | 77 | |
AK295905.1 | - | 19 - 20 | 2174 | 77 | |
AK296258.1 | - | 19 - 20 | 2103 | 77 | |
AL096717.1 | - | 16 - 17 | 1606 | 77 | |
BC032630.1 | - | 14 - 15 | 1454 | 77 |