The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193268.1 | NP_001180197.1 | 7 - 8 | 1005 | 71 |
NM_001193269.1 | NP_001180198.1 | 7 - 8 | 909 | 71 | |
NM_012155.2 | NP_036287.1 | 4 - 5 | 370 | 71 | |
XM_006723125.2 | XP_006723188.1 | 7 - 8 | 1086 | 71 | |
XM_006723126.2 | XP_006723189.1 | 7 - 8 | 894 | 71 | |
XM_011526694.2 | XP_011524996.1 | 7 - 8 | 1089 | 71 | |
XM_011526695.2 | XP_011524997.1 | 7 - 8 | 1089 | 71 | |
XM_011526698.2 | XP_011525000.1 | 4 - 5 | 412 | 71 | |
XM_017026550.1 | XP_016882039.1 | 4 - 5 | 350 | 71 | |
XM_017026551.1 | XP_016882040.1 | 7 - 8 | 1089 | 71 | |
GenBank mRNA | AB209773.1 | - | 1 - 2 | 106 | 71 |
AF103939.1 | - | 4 - 5 | 364 | 71 | |
AK225043.1 | - | 4 - 5 | 356 | 71 | |
AK294668.1 | - | 7 - 8 | 956 | 71 | |
AK295905.1 | - | 7 - 8 | 909 | 71 | |
AK296258.1 | - | 7 - 8 | 838 | 71 | |
AK302961.1 | - | 7 - 8 | 969 | 71 | |
AK304299.1 | - | 4 - 5 | 354 | 71 | |
AK308403.1 | - | 4 - 5 | 350 | 71 | |
AL096717.1 | - | 4 - 5 | 342 | 71 | |
BC029304.1 | - | 7 - 8 | 1072 | 71 | |
BC032630.1 | - | 4 - 5 | 355 | 71 | |
DC366776.1 | - | 4 - 5 | 370 | 71 |