The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243027.2 | NP_001229956.1 | 4 - 5 | 548 | 70 |
NM_001243028.2 | NP_001229957.1 | 3 - 4 | 455 | 70 | |
NM_001626.5 | NP_001617.1 | 4 - 5 | 585 | 70 | |
XM_011526614.1 | XP_011524916.1 | 4 - 5 | 388 | 70 | |
XM_011526615.1 | XP_011524917.1 | 4 - 5 | 465 | 70 | |
XM_011526616.1 | XP_011524918.1 | 4 - 5 | 708 | 70 | |
XM_011526618.1 | XP_011524920.1 | 4 - 5 | 550 | 70 | |
XM_011526619.1 | XP_011524921.1 | 4 - 5 | 519 | 70 | |
XM_011526620.1 | XP_011524922.1 | 4 - 5 | 454 | 70 | |
XM_011526622.2 | XP_011524924.1 | 4 - 5 | 532 | 70 | |
XM_017026470.1 | XP_016881959.1 | 5 - 6 | 667 | 70 | |
XM_017026471.1 | XP_016881960.1 | 4 - 5 | 532 | 70 | |
GenBank mRNA | AK294453.1 | - | 4 - 5 | 495 | 70 |
AK304209.1 | - | 4 - 5 | 594 | 70 | |
AK308006.1 | - | 3 - 4 | 325 | 70 | |
AK308906.1 | - | 3 - 4 | 402 | 70 | |
AK314619.1 | - | 4 - 5 | 527 | 70 | |
BC032709.1 | - | 4 - 5 | 439 | 70 | |
BC040028.1 | - | 4 - 5 | 550 | 70 | |
BC063421.1 | - | 4 - 5 | 577 | 70 | |
BC120994.1 | - | 3 - 4 | 331 | 70 | |
BC120995.1 | - | 3 - 4 | 331 | 70 | |
BQ219902.1 | - | 4 - 5 | 548 | 70 | |
KU159425.1 | - | 4 - 5 | 532 | 70 | |
M77198.1 | - | 4 - 5 | 489 | 70 | |
M95936.1 | - | 3 - 4 | 373 | 70 |