The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007464.2 | NP_001007465.1 | 7 - 8 | 127 | |
NM_015952.3 | NP_057036.2 | 6 - 7 | 127 | ||
NM_016104.3 | NP_057188.2 | 8 - 9 | 127 | ||
GenBank mRNA | AF092134.1 | - | 6 - 7 | 127 | |
AF132958.1 | - | 6 - 7 | 127 | ||
AI129713.1 | - | 1 - 2 | 293 | 127 | |
AK290727.1 | - | 6 - 7 | 127 | ||
AK301089.1 | - | 7 - 8 | 127 | ||
BC015802.1 | - | 6 - 7 | 127 | ||
BQ001584.1 | - | 1 - 2 | 309 | 127 | |
CA422968.1 | - | 1 - 2 | 305 | 127 |