The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024100.3 | NP_077005.2 | 4 - 5 | 622 | 128 |
XM_017027027.1 | XP_016882516.1 | 6 - 7 | 1148 | 128 | |
GenBank mRNA | BC000040.2 | - | 4 - 5 | 607 | 128 |
BC001648.2 | - | 4 - 5 | 602 | 128 |