The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005727.3 | NP_005718.2 | 4 - 5 | 740 | 128 |
XM_011540460.2 | XP_011538762.1 | 4 - 5 | 714 | 128 | |
GenBank mRNA | AF054838.1 | - | 2 - 3 | 266 | 128 |
AF065388.1 | - | 3 - 4 | 387 | 128 | |
AF133425.1 | - | 3 - 4 | 389 | 128 | |
AK313774.1 | - | 4 - 5 | 730 | 128 | |
BC007290.1 | - | 3 - 4 | 393 | 128 | |
BC013404.2 | - | 3 - 4 | 391 | 128 |