Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNAJC11
Entrez Gene ID
55735
Gene Name
DnaJ heat shock protein family (Hsp40) member C11
Gene Aliases
dJ126A5.1
Chromosome Location
Chr.1: 6634168 - 6701906 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018198.3 NP_060668.2 13 - 14 1492 66
XM_017001745.1 XP_016857234.1 12 - 13 1345 66
GenBank mRNA AF306695.1 - 13 - 14 1406 66
AK001599.1 - 13 - 14 1401 66
AK025491.1 - 3 - 4 166 66
AK093709.1 - 1 - 2 647 66
AK095386.1 - 4 - 5 1214 66
AK294539.1 - 12 - 13 1315 66
AK298378.1 - 13 - 14 1378 66
AL833841.1 - 13 - 14 1366 66
BC006086.1 - 13 - 14 1388 66
BC008772.1 - 13 - 14 1393 66
BC014145.2 - 12 - 13 1244 66

View Full Product Details