Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJC11
Entrez Gene ID
55735
Gene Name
DnaJ heat shock protein family (Hsp40) member C11
Gene Aliases
dJ126A5.1
Chromosome Location
Chr.1: 6634168 - 6701906 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018198.3 NP_060668.2 5 - 6 612 92
XM_017001745.1 XP_016857234.1 4 - 5 465 92
GenBank mRNA AF306694.1 - 5 - 6 511 92
AF306695.1 - 5 - 6 526 92
AF306696.1 - 3 - 4 316 92
AK001599.1 - 5 - 6 521 92
AK294539.1 - 4 - 5 435 92
AK298378.1 - 5 - 6 498 92
AL109978.1 - 4 - 5 434 92
AL833841.1 - 5 - 6 486 92
AY336749.1 - 5 - 6 522 92
BC006086.1 - 5 - 6 508 92
BC008772.1 - 5 - 6 513 92
BC014145.2 - 5 - 6 520 92

View Full Product Details