Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJC11
Entrez Gene ID
55735
Gene Name
DnaJ heat shock protein family (Hsp40) member C11
Gene Aliases
dJ126A5.1
Chromosome Location
Chr.1: 6634168 - 6701906 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018198.3 NP_060668.2 6 - 7 741 72
XM_017001745.1 XP_016857234.1 5 - 6 594 72
GenBank mRNA AF306695.1 - 6 - 7 655 72
AK001599.1 - 6 - 7 650 72
AK294539.1 - 5 - 6 564 72
AK298378.1 - 6 - 7 627 72
AL109978.1 - 5 - 6 563 72
AL833841.1 - 6 - 7 615 72
AY336749.1 - 6 - 7 651 72
BC006086.1 - 6 - 7 637 72
BC008772.1 - 6 - 7 642 72
BC014145.2 - 6 - 7 649 72

View Full Product Details