Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZMYND12
Entrez Gene ID
84217
Gene Name
zinc finger MYND-type containing 12
Gene Aliases
-
Chromosome Location
Chr.1: 42430329 - 42456267 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032257.4 NP_115633.3 3 - 4 698 121
GenBank mRNA BC024186.2 - 3 - 4 687 121

View Full Product Details