Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZMYND12
Entrez Gene ID
84217
Gene Name
zinc finger MYND-type containing 12
Gene Aliases
-
Chromosome Location
Chr.1: 42430329 - 42456267 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146192.1 NP_001139664.1 5 - 6 924 90
NM_032257.4 NP_115633.3 6 - 7 1096 90
GenBank mRNA AK057384.1 - 6 - 7 1096 90
AK301839.1 - 5 - 6 910 90
AL136858.1 - 6 - 7 903 90
BC024186.2 - 6 - 7 1085 90
BQ441452.1 - 3 - 4 385 90

View Full Product Details