The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308220.1 | NP_001295149.2 | 8 - 9 | 1168 | 65 |
NM_033266.3 | NP_150296.4 | 8 - 9 | 1168 | 65 | |
XM_011545708.1 | XP_011544010.1 | 9 - 10 | 994 | 65 | |
XM_011545709.1 | XP_011544011.1 | 10 - 11 | 1045 | 65 | |
XM_011545711.2 | XP_011544013.1 | 8 - 9 | 916 | 65 | |
XM_011545712.2 | XP_011544014.1 | 8 - 9 | 916 | 65 | |
XM_011545713.1 | XP_011544015.1 | 8 - 9 | 916 | 65 | |
GenBank mRNA | AK131280.1 | - | 8 - 9 | 1168 | 65 |
AK310834.1 | - | 9 - 10 | 990 | 65 | |
EF560720.1 | - | 8 - 9 | 895 | 65 | |
EF560739.1 | - | 8 - 9 | 899 | 65 |