Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
XPO7
Entrez Gene ID
23039
Gene Name
exportin 7
Gene Aliases
EXP7, RANBP16
Chromosome Location
Chr.8: 21919669 - 22006585 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015024.4 NP_055839.3 2 - 3 269 81
XM_017013244.1 XP_016868733.1 2 - 3 496 81
XM_017013245.1 XP_016868734.1 2 - 3 263 81
XM_017013246.1 XP_016868735.1 2 - 3 492 81
XM_017013247.1 XP_016868736.1 2 - 3 656 81
GenBank mRNA AB018288.1 - 2 - 3 179 81
AF064729.1 - 2 - 3 179 81
AK291472.1 - 2 - 3 184 81
AK311616.1 - 2 - 3 362 81
BC030785.1 - 2 - 3 268 81
DB460798.1 - 2 - 3 271 81
DC332379.1 - 2 - 3 191 81

View Full Product Details