Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
XPO7
Entrez Gene ID
23039
Gene Name
exportin 7
Gene Aliases
EXP7, RANBP16
Chromosome Location
Chr.8: 21919669 - 22006585 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015024.4 NP_055839.3 21 - 22 2447 88
XM_017013244.1 XP_016868733.1 21 - 22 2701 88
XM_017013245.1 XP_016868734.1 21 - 22 2468 88
XM_017013246.1 XP_016868735.1 21 - 22 2670 88
GenBank mRNA AB018288.1 - 18 - 19 1811 88
AF064729.1 - 21 - 22 2357 88
AK291472.1 - 21 - 22 2362 88
AK299848.1 - 17 - 18 1883 88
BC030785.1 - 21 - 22 2446 88

View Full Product Details