Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
XPO7
Entrez Gene ID
23039
Gene Name
exportin 7
Gene Aliases
EXP7, RANBP16
Chromosome Location
Chr.8: 21919669 - 22006585 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015024.4 NP_055839.3 25 - 26 3039 110
XM_017013244.1 XP_016868733.1 25 - 26 3293 110
XM_017013245.1 XP_016868734.1 25 - 26 3060 110
XM_017013246.1 XP_016868735.1 25 - 26 3262 110
GenBank mRNA AB018288.1 - 22 - 23 2403 110
AF064729.1 - 25 - 26 2949 110
AK291472.1 - 25 - 26 2954 110
AK299848.1 - 21 - 22 2475 110
BC014219.2 - 2 - 3 298 110
BC030785.1 - 25 - 26 3038 110

View Full Product Details