Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XPO7
Entrez Gene ID
23039
Gene Name
exportin 7
Gene Aliases
EXP7, RANBP16
Chromosome Location
Chr.8: 21919669 - 22006585 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015024.4 NP_055839.3 3 - 4 363 62
XM_017013244.1 XP_016868733.1 3 - 4 590 62
XM_017013245.1 XP_016868734.1 3 - 4 357 62
XM_017013246.1 XP_016868735.1 3 - 4 586 62
XM_017013247.1 XP_016868736.1 3 - 4 750 62
GenBank mRNA AB018288.1 - 3 - 4 273 62
AF064729.1 - 3 - 4 273 62
AK291472.1 - 3 - 4 278 62
AK311616.1 - 3 - 4 456 62
AL597247.1 - 3 - 4 319 62
BC030785.1 - 3 - 4 362 62
DC332379.1 - 3 - 4 285 62

View Full Product Details