The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015004.3 | NP_055819.2 | 6 - 7 | 648 | 60 |
NR_023353.1 | 4 - 5 | 451 | 60 | ||
XM_017005930.1 | XP_016861419.1 | 4 - 5 | 466 | 60 | |
GenBank mRNA | AK090511.1 | - | 1 - 2 | 1626 | 60 |
AK130053.1 | - | 6 - 7 | 792 | 60 | |
AK315741.1 | - | 6 - 7 | 648 | 60 | |
BC012831.2 | - | 6 - 7 | 625 | 60 | |
BU153856.1 | - | 4 - 5 | 459 | 60 | |
D29958.1 | - | 6 - 7 | 617 | 60 |