The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015004.3 | NP_055819.2 | 7 - 8 | 801 | 72 |
NR_023353.1 | 5 - 6 | 604 | 72 | ||
XM_017005930.1 | XP_016861419.1 | 5 - 6 | 619 | 72 | |
GenBank mRNA | AK130053.1 | - | 7 - 8 | 945 | 72 |
AK315741.1 | - | 7 - 8 | 801 | 72 | |
BC012831.2 | - | 7 - 8 | 778 | 72 | |
D29958.1 | - | 7 - 8 | 770 | 72 |