The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001447.2 | NP_001438.1 | 14 - 15 | 10038 | 114 |
XM_006714761.3 | XP_006714824.1 | 15 - 16 | 10609 | 114 | |
XM_011537600.2 | XP_011535902.1 | 17 - 18 | 10377 | 114 | |
XM_011537603.2 | XP_011535905.1 | 15 - 16 | 10310 | 114 | |
XM_017009224.1 | XP_016864713.1 | 17 - 18 | 10379 | 114 | |
XM_017009225.1 | XP_016864714.1 | 16 - 17 | 10221 | 114 | |
GenBank mRNA | AB160984.1 | - | 2 - 3 | 606 | 114 |
AF231022.1 | - | 14 - 15 | 10038 | 114 |