The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001447.2 | NP_001438.1 | 17 - 18 | 10519 | 102 |
XM_006714761.3 | XP_006714824.1 | 18 - 19 | 11090 | 102 | |
XM_011537600.2 | XP_011535902.1 | 20 - 21 | 10858 | 102 | |
XM_011537603.2 | XP_011535905.1 | 18 - 19 | 10791 | 102 | |
XM_017009224.1 | XP_016864713.1 | 20 - 21 | 10860 | 102 | |
XM_017009225.1 | XP_016864714.1 | 19 - 20 | 10702 | 102 | |
GenBank mRNA | AB160984.1 | - | 5 - 6 | 1087 | 102 |
AF231022.1 | - | 17 - 18 | 10519 | 102 |