Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OMP
Entrez Gene ID
4975
Gene Name
olfactory marker protein
Gene Aliases
-
Chromosome Location
Chr.11: 77102840 - 77103331 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006189.1 NP_006180.1 1 - 1 342 57
GenBank mRNA BC069115.1 - 1 - 1 342 57
BC069365.1 - 1 - 1 342 57

View Full Product Details