Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPR65
Entrez Gene ID
8477
Gene Name
G protein-coupled receptor 65
Gene Aliases
TDAG8, hTDAG8
Chromosome Location
Chr.14: 88005124 - 88014811 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK313797.1 - 1 - 2 70 86
BC035633.1 - 1 - 2 67 86

View Full Product Details