The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284266.1 | NP_001271195.1 | 3 - 4 | 330 | 124 |
NM_001284267.1 | NP_001271196.1 | 3 - 4 | 183 | 124 | |
NM_001284268.1 | NP_001271197.1 | 3 - 4 | 341 | 124 | |
NM_001284269.1 | NP_001271198.1 | 3 - 4 | 341 | 124 | |
NM_145231.3 | NP_660274.1 | 3 - 4 | 330 | 124 | |
GenBank mRNA | AK094740.1 | - | 3 - 4 | 283 | 124 |
AK298930.1 | - | 3 - 4 | 275 | 124 | |
AL832321.1 | - | 3 - 4 | 267 | 124 | |
BC001787.1 | - | 3 - 4 | 242 | 124 | |
BG494945.1 | - | 3 - 4 | 295 | 124 | |
BP371056.1 | - | 3 - 4 | 183 | 124 | |
BX161414.1 | - | 3 - 4 | 307 | 124 | |
CF593330.1 | - | 3 - 4 | 326 | 124 |