Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFCAB11
Entrez Gene ID
90141
Gene Name
EF-hand calcium binding domain 11
Gene Aliases
C14orf143
Chromosome Location
Chr.14: 89794984 - 89954777 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284266.1 NP_001271195.1 4 - 5 440 122
NM_001284267.1 NP_001271196.1 4 - 5 293 122
NM_001284268.1 NP_001271197.1 4 - 5 451 122
NM_001284269.1 NP_001271198.1 4 - 5 451 122
NM_145231.3 NP_660274.1 4 - 5 440 122
GenBank mRNA AK094740.1 - 4 - 5 393 122
AK298930.1 - 4 - 5 385 122
AK310331.1 - 3 - 4 352 122
AL832321.1 - 4 - 5 377 122
BC001787.1 - 4 - 5 352 122
BG494945.1 - 4 - 5 405 122
BP371056.1 - 4 - 5 293 122
BX161414.1 - 4 - 5 417 122
BX247991.1 - 4 - 5 358 122
CF593330.1 - 4 - 5 436 122

View Full Product Details