Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC4A
Entrez Gene ID
50856
Gene Name
C-type lectin domain family 4 member A
Gene Aliases
CD367, CLECSF6, DCIR, DDB27, HDCGC13P, LLIR
Chromosome Location
Chr.12: 8102932 - 8138607 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016184.3 NP_057268.1 3 - 4 546 133
NM_194447.2 NP_919429.2 2 - 3 429 133
XM_011520684.2 XP_011518986.1 2 - 3 1807 133
XM_017019382.1 XP_016874871.1 4 - 5 467 133
GenBank mRNA AF067800.1 - 3 - 4 567 133
AF109146.1 - 3 - 4 393 133
AF328684.1 - 3 - 4 538 133
AJ133532.1 - 3 - 4 540 133
BC074822.2 - 3 - 4 321 133
BC074874.2 - 3 - 4 321 133
BC117439.1 - 3 - 4 441 133
BC117441.1 - 3 - 4 441 133

View Full Product Details