The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287804.1 | NP_001274733.1 | 1 - 2 | 219 | 115 |
NM_032327.3 | NP_115703.2 | 1 - 2 | 219 | 115 | |
NM_198043.2 | NP_932160.1 | 1 - 2 | 219 | 115 | |
NM_198044.2 | NP_932161.1 | 1 - 2 | 219 | 115 | |
NM_198045.2 | NP_932162.1 | 1 - 2 | 219 | 115 | |
XM_006718021.1 | XP_006718084.1 | 1 - 2 | 207 | 115 | |
XM_006718023.1 | XP_006718086.1 | 1 - 2 | 207 | 115 | |
XM_006718024.1 | XP_006718087.1 | 1 - 2 | 207 | 115 | |
XM_006718025.1 | XP_006718088.1 | 1 - 2 | 207 | 115 | |
XM_006718026.1 | XP_006718089.1 | 1 - 2 | 207 | 115 | |
XM_017016767.1 | XP_016872256.1 | 1 - 2 | 207 | 115 | |
XM_017016769.1 | XP_016872258.1 | 1 - 2 | 207 | 115 | |
XM_017016770.1 | XP_016872259.1 | 1 - 2 | 172 | 115 | |
XM_017016772.1 | XP_016872261.1 | 1 - 2 | 172 | 115 | |
GenBank mRNA | AF258563.1 | - | 1 - 2 | 148 | 115 |
AK074427.1 | - | 1 - 2 | 173 | 115 | |
AY359080.1 | - | 1 - 2 | 110 | 115 | |
BC004535.2 | - | 1 - 2 | 140 | 115 | |
BC008074.2 | - | 1 - 2 | 177 | 115 | |
BC011749.2 | - | 1 - 2 | 165 | 115 | |
BC012830.2 | - | 1 - 2 | 177 | 115 | |
BC033157.1 | - | 1 - 2 | 146 | 115 | |
BX353685.2 | - | 1 - 2 | 156 | 114 |