Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ZDHHC16
- Entrez Gene ID
-
84287
- Gene Name
- zinc finger DHHC-type containing 16
- Gene Aliases
-
APH2, DHHC-16
- Chromosome Location
-
Chr.10: 97446131 - 97457370 on Build GRCh38
View Full Product Details