The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005439.2 | NP_005430.1 | 3 - 4 | 244 | 103 |
NR_026581.1 | 3 - 4 | 741 | 103 | ||
GenBank mRNA | AF070539.1 | - | 3 - 4 | 374 | 103 |
AK289869.1 | - | 3 - 4 | 244 | 103 | |
AK313998.1 | - | 3 - 4 | 545 | 103 | |
BC000898.2 | - | 3 - 4 | 314 | 103 | |
BC002340.2 | - | 3 - 4 | 354 | 103 | |
BT019393.1 | - | 2 - 3 | 181 | 103 | |
BT019432.1 | - | 2 - 3 | 181 | 103 | |
CR541700.1 | - | 2 - 3 | 181 | 103 | |
U57342.1 | - | 3 - 4 | 360 | 103 |